  • Search Results
  • Cardiomyocyte lipotoxicity is mediated by Il-6 and causes down-regulation of PPARs.
Cardiomyocyte lipotoxicity is mediated by Il-6 and causes down-regulation of PPARs.
Biochemical and biophysical research communications (2015-02-26)
Taha Haffar, Félix-Antoine Bérubé-Simard, Nicolas Bousette
ABSTRACT
Here we sought to evaluate the effect of palmitate on cytokine and PPAR activity/expression. We investigated the effect of BSA conjugated palmitate and oleate on PPAR activity, PPAR-α and δ expression, as well as the expression of cytokines and key factors responsible for β-oxidation by qRT-PCR and western blotting in primary rat neonatal cardiomyocytes (NCMs). Furthermore we evaluated the effect of anti-inflammatory actions of AICAR and PPAR agonists on cytokine expression and cell death in palmitate treated NCMs. We found that palmitate caused down regulation of PPARs and increased cytokine expression and cell death, all of which was significantly attenuated by the co-administration of either AICAR or PPAR agonists. This work supports the pro-inflammatory actions of intracellular lipid and provides further insight into the pathological mechanism of cardiac lipotoxicity as occurs in diabetic hearts.
Product Description
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)