  • Search Results
  • Sulfonylurea induction of caffeine-enhanced insulin secretion and reduction of glycemic levels in diabetic rats.
Sulfonylurea induction of caffeine-enhanced insulin secretion and reduction of glycemic levels in diabetic rats.
Pharmaceutical biology (2014-03-08)
Luiz Augusto da Silva, Ricardo Aparecido Pereira, Janaína Angela Túrmina, Ivo Ilvan Kerppers, Raul Osiecki, Leandro Ricardo Altimari, Carlos Ricardo Maneck Malfatti
ABSTRACT
Caffeine can stimulate insulin secretion by attenuating hyperglycemia in diabetes models with significant reduction of pancreatic functional β cells. Knowledge of these mechanisms could contribute to new strategies for treating diabetes. This study evaluated the effects of caffeine and physical exercise on glycemic and insulin responses in diabetic rats. The diabetes model was induced by intraperitoneal administration of 60 mg/kg of streptozotocin (STZ). Animals were divided into six groups: control, caffeine, STZ control, STZ caffeine, STZ sulfonylurea, and STZ caffeine + sulfonylurea. Acutely, control animals received 6 mg of caffeine and 10 mg/kg sulfonylurea or 10 mg/kg saline. Animals were sacrificed after physical exercise; blood samples were collected for glucose, glycerol, lactate, and insulin analyses. Cardiovascular responses were recorded before and after treatments. A one-way ANOVA and the post hoc Student-Newman-Keuls test were used to analyze statistical differences between treatments (p < 0.05). About 6 mg/kg of caffeine did not alter cardiovascular responses, but promoted blood glucose reduction after 60 min of exercise when compared to animals in the control groups (387-187 mg/dL; p < 0.05). Insulin levels increased significantly (0.6-10 µU/mL; p < 0.05) in rats that received acute caffeine treatment associated with sulfonylurea compared to rats in the other groups. Acute caffeine intake with exercise can increase glucose uptake enhancing insulin secretion stimulated by sulfonylurea in β cells-deficient pancreas. The results indicate the potential use of caffeine as a strategy for glycemic and insulin control in diabetes.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard