  • Search Results
  • Lymphangiogenesis in gastric cancer regulated through Akt/mTOR-VEGF-C/VEGF-D axis.
Lymphangiogenesis in gastric cancer regulated through Akt/mTOR-VEGF-C/VEGF-D axis.
BMC cancer (2015-04-18)
Hongxia Chen, Runnian Guan, Yupeng Lei, Jianyong Chen, Qi Ge, Xiaoshen Zhang, Ruoxu Dou, Hongyuan Chen, Hao Liu, Xiaolong Qi, Xiaodong Zhou, Changyan Chen
ABSTRACT
Lymphangiogenesis plays a significant role in metastasis and recurrence of gastric cancer. There is no report yet focusing on the modulation of VEGF pathway and lymphangiogenesis of gastric cancer by targeting Akt/mTOR pathway. This study aims to demonstrate the relationship between Akt/mTOR pathway and VEGF-C/-D in gastric cancer. We collected surgically resected gastric adenocarcinoma specimens from 55 consented patients. Immunohistochemistry staining of p-Akt, p-mTOR, VEGF-C, VEGF-D were performed and scored by two independent pathologists. The results were presented as staining intensity and positive staining cell rate. We also measured lymphatic vessel density (LVD) by D2-40 staining. Different dosages of p-Akt inhibitor LY294002 (12.5 μM, 25 μM, 50 μM) and p-mTOR inhibitor Rapamycin (25 nM, 50 nM, 100 nM) were given to gastric cancer cell line SGC-7901 in vitro. The inhibition rate of cell growth was tested by MTT at 24 h, 48 h and 72 h, respectively and protein expressions of Akt, p-Akt, mTOR, p-mTOR, VEGF-C and VEGF-D were examined by Western blot. The positive staining rates of p-Akt, p-mTOR, VEGF-C and VEGF-D in 55 gastric cancer clinical specimens were 74.54%, 85.45%, 72.73% and 58.18%. p-Akt and p-mTOR were positively correlated with VEGF-C and VEGF-D (p < 0.01). The LVD increased with incremental tendency of staining intensity of p-Akt, p-mTOR, VEGF-C and VEGF-D. LY294002 or Rapamycin significantly suppressed SGC-7901 cell growth and the inhibition rate was dose and time dependent (p < 0.001). In addition, the protein expression of p-Akt and p-mTOR were positively correlated with that of VEGF-C and VEGF-D (p < 0.05). The level of LVD in gastric cancer specimens was significant higher than that of normal gastric tissue and was positively correlated with p-Akt, p-mTOR, VEGF-C and VEGF-D. Inhibition of p-Akt and p-mTOR, in vitro, decreased tumor cell VEGF-C and VEGF-D significantly. Therefore, we concluded that lymphangiogenesis of gastric cancer might be related to Akt/mTOR-VEGF-C/VEGF-D axis.
Sigma-Aldrich
Glycerol solution, puriss. p.a., 86-89% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M