  • Search Results
  • A novel UV degradation product of Ebastine: isolation and characterization using Q-TOF, NMR, IR and computational chemistry.
A novel UV degradation product of Ebastine: isolation and characterization using Q-TOF, NMR, IR and computational chemistry.
Journal of pharmaceutical and biomedical analysis (2015-02-14)
Ravi Rapolu, Avadhesh Kumar Pandey, Ch Krishnam Raju, Kaushik Ghosh, Kolupula Srinivas, Atul Awasthi, Sameer G Navalgund, Koduru V Surendranath
ABSTRACT
Forced degradation of Ebastine (1-(4-(1,1-dimethylethyl)phenyl)-4-(4-(diphenylmethoxy) piperidin-1-yl)butan-1-one) drug substance in ultraviolet light condition resulted into an unknown significant degradation product. This degradation product was analyzed using a newly developed reverse-phase HPLC, where it was eluted at 2.73 relative retention time to Ebastine peak. UV degradation product was isolated from reaction mass using preparative HPLC and its structure was elucidated using high resolution MS, multidimensional NMR and FTIR spectroscopic techniques. UV degradation product has been characterized as 2-(4-(benzhydryloxy)piperidin-1-yl)-1-(4-(tert-butyl)phenyl)-2-methylcyclopropanol. (1)H and (13)C NMR chemical shift values were generated using computational chemistry for possible two diastereomers (7R10S and 7R10R) and later 7R10R was confirmed (and its enantiomer) as final structure given it showed close agreement with experimental NMR data. Formation of UV degradation product as a recemic mixture was further verified by computational chemistry evaluation, chiral HPLC and polarimetery. To best of our knowledge, this is a novel degradation product which is not discussed at any form of publication yet.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Diethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Diethylamine, BioXtra
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Diethylamine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Diethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%