  • Search Results
  • Tissue factor and Toll-like receptor (TLR)4 in hyperglycaemia-hyperinsulinaemia. Effects in healthy subjects, and type 1 and type 2 diabetes mellitus.
Tissue factor and Toll-like receptor (TLR)4 in hyperglycaemia-hyperinsulinaemia. Effects in healthy subjects, and type 1 and type 2 diabetes mellitus.
Thrombosis and haemostasis (2015-02-06)
Anamika Singh, Guenther Boden, A Koneti Rao
ABSTRACT
Diabetes mellitus (DM) patients have an increased incidence of cardiovascular events. Blood tissue factor-procoagulant activity (TF-PCA), the initiating mechanism for blood coagulation, is elevated in DM. We have shown that hyperglycaemia (HG), hyperinsulinaemia (HI) and combined HG+HI (induced using 24-hour infusion clamps) increases TF-PCA in healthy and type 2 DM (T2DM) subjects, but not in type 1 DM (T1DM) subjects. The mechanisms for this are unknown. DM patients have elevated plasma lipopolysaccharide (LPS), a toll-like receptor (TLR) 4 ligand. We postulated that TLR4 plays a role in modulating TF levels. We studied the effect of HG+HI on TLR4 and TF-PCA in vivo during 24-hour HG+HI infusion clamps in healthy subjects, and T1DM and T2DM subjects, and in vitro in blood. In vivo, in healthy subjects, 24-hour HG + HI infusion increased TLR4 six-fold, which correlated with TF-PCA (r= 0.91, p<0.0001). T2DM patients showed smaller increases in both. In T1DM subjects, TLR4 declined (50%, p<0.05) and correlated with TF-PCA (r=0.55; p<0.05). In vitro, HG (200 mg/dl added glucose) and HI (1-100 nM added insulin) increased TF-PCA in healthy subjects (~2-fold, 2-4 hours). Insulin inhibited by ~30% LPS-induced increase in TF-PCA and high glucose reversed it. TLR4 levels paralleled TF-PCA (r=0.71, p<0.0001); HG and HI increased TLR4 and insulin inhibited LPS-induced TLR4 increase. This is first evidence that even in healthy subjects, HG of short duration increases TLR4 and TF-PCA, key players in inflammation and thrombosis. TLR4-TF interplay is strikingly different in non-diabetic, T1DM and T2DM subjects.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, AR, anhydrous