  • Search Results
  • Plasma sphingolipids as potential indicators of hepatic necroinflammation in patients with chronic hepatitis C and normal alanine aminotransferase level.
Plasma sphingolipids as potential indicators of hepatic necroinflammation in patients with chronic hepatitis C and normal alanine aminotransferase level.
PloS one (2014-04-17)
Jun-Feng Li, Feng Qu, Su-Jun Zheng, Jin-Yu Ren, Hui-Li Wu, Mei Liu, Hui Liu, Feng Ren, Yu Chen, Jin-Lan Zhang, Zhong-Ping Duan
ABSTRACT
Accurate estimation of hepatic necroinflammation caused by chronic hepatitis C (CHC) is crucial for prediction of prognosis and design of therapeutic strategy, which is particularly true for CHC patients with normal alanine aminotransferase (ALT) level. Recent studies have shown that sphingolipids have a close relationship with hepatitis C virus infection. The present study aimed to identify plasma sphingolipids related to hepatic necroinflammation. We included 120 treatment-naïve CHC patients and 64/120 had normal ALT levels (<40 U/L). CHC patients who underwent liver biopsies were subjected to Scheuer scoring analysis for scope of hepatic inflammation. Plasma sphingolipids were detected by high-performance liquid chromatography tandem mass spectrometry. Our results showed 44 plasma sphingolipids were detected altogether. Of all detected sphingolipids, hexosylceramide (HexCer) (d18∶1/22∶0) and HexCer (d18∶1/24∶0) showed a significant difference among G0/G1, G2, and G3/G4 (P<0.05). For identifying hepatic necroinflammation (G≥2), after adjusting other factors, the odds ratio (OR) of HexCer (d18∶1/22∶0) reached 1.01 (95% confidence interval [CI]: 1.00-1.02). Furthermore, the area under the curve (AUC) of HexCer (d18∶1/22∶0) was 0.7 (P = 0.01) and approached that of ALT (AUC = 0.78). However, in CHC patients with normal ALT, HexCer (d18∶1/22∶0) was an independent factor (OR: 1.02, 95% CI: 1.01-1.03) to identify the hepatic necroinflammation (G≥2). HexCer (d18∶1/22∶0) not only showed the largest AUC (0.78, P = 0.001), but also exhibited the highest specificity of all indicators. These results indicate that plasma HexCer (d18∶1/22∶0) is a potential indicator to distinguish hepatic necroinflammation in CHC patients. For CHC with normal ALT, the ability of HexCer (d18∶1/22∶0) to distinguish hepatic necroinflammation might be superior to conventional serum indicators.
Product Description
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis