  • Search Results
  • Synthesis and evaluation of anticonvulsant activity of N-(2,5-dimethylphenoxy)- and N-[(2,3,5-trimethylphenoxy)alkyl]aminoalkanols.
Synthesis and evaluation of anticonvulsant activity of N-(2,5-dimethylphenoxy)- and N-[(2,3,5-trimethylphenoxy)alkyl]aminoalkanols.
Acta poloniae pharmaceutica (2015-04-09)
Anna Maria Waszkielewicz, Agnieszka Gunia-Krzyżak, Marek Cegła, Henryk Marona
ABSTRACT
A series of new N-(2,5-dimethylphenoxy)- and N-(2,3,5-trimethylphenoxy)alkylaminoalkanols [I-XVII] was synthesized and evaluated for anticonvulsant activity. Pharmacological tests included maximal electroshock (MES) and subcutaneous pentetrazole seizure threshold (scMet) assays as well as neurotoxicity (TOX) evaluation in mice after intraperitoneal (i.p.) administration and/or in rats after oral (p.o.) administration. The most active compound was R-2N-[(2,3,5-trimethylphenoxy)ethyl]aminobutan-1-ol, which exhibited 100% activity in MES at the dose of 30 mg/kg body weight (mice, i.p.) and 75% activity in MES at 30 mg/kg b.w. (rats, p.o.) without neurotoxicity at the active doses.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%