  • Search Results
  • Absolute quantification of podocin, a potential biomarker of glomerular injury in human urine, by liquid chromatography-multiple reaction monitoring cubed mass spectrometry.
Absolute quantification of podocin, a potential biomarker of glomerular injury in human urine, by liquid chromatography-multiple reaction monitoring cubed mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-02-21)
Romain Simon, Jérôme Lemoine, Catherine Fonbonne, Aurore Jaffuel, Jean-François Léonard, Jean-Charles Gautier, Olivier Pasquier, Arnaud Salvador
ABSTRACT
Glomeruli play a major role in the kidney function since they are involved in primary urine formation. It is then crucial to dispose of methods to monitor glomerular injury, especially in drug development. In this context, quantification of podocin could be of great interest since it is a protein exclusively present in highly specialized glomerulus cells called podocytes. Immunoassays are the most commonly used approach for protein assays. However, they rely on the availability of specific antibodies. When such antibodies are not available, liquid chromatography tandem mass spectrometry (LC-MS/MS), in selected reaction monitoring (SRM) or in multiple reaction monitoring cubed (MRM(3)) mode, has been demonstrated as a powerful alternative technique, and can be applied to multiple protein quantification. This paper describes the development of a quantification method of human podocin in urine by LC-MS/MS in MRM(3) mode. Inter assay precision and accuracy ranged from 7 to 20% and from 105 to 112% respectively and the lower limit of quantification (LLOQ) was 0.39ng/mL from only 1mL of urine which is compatible for endogenous level of podocin determination.
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Urea, European Pharmacopoeia (EP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urea, analytical standard
SAFC
Sodium deoxycholate
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Formic acid, SAJ first grade, 88.0-89.5%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000