  • Search Results
  • Fast dissolving paracetamol/caffeine nanofibers prepared by electrospinning.
Fast dissolving paracetamol/caffeine nanofibers prepared by electrospinning.
International journal of pharmaceutics (2014-12-03)
U Eranka Illangakoon, Hardyal Gill, Gemma C Shearman, Maryam Parhizkar, Sunthar Mahalingam, Nicholas P Chatterton, Gareth R Williams
ABSTRACT
A series of polyvinylpyrrolidone fibers loaded with paracetamol (PCM) and caffeine (CAF) was fabricated by electrospinning and explored as potential oral fast-dissolving films. The fibers take the form of uniform cylinders with smooth surfaces, and contain the drugs in the amorphous form. Drug-polymer intermolecular interactions were evidenced by infrared spectroscopy and molecular modeling. The properties of the fiber mats were found to be highly appropriate for the preparation of oral fast dissolving films: their thickness is around 120-130 μm, and the pH after dissolution in deionized water lies in the range of 6.7-7.2. Except at the highest drug loading, the folding endurance of the fibers was found to be >20 times. A flavoring agent can easily be incorporated into the formulation. The fiber mats are all seen to disintegrate completely within 0.5s when added to simulated saliva solution. They release their drug cargo within around 150s in a dissolution test, and to undergo much more rapid dissolution than is seen for the pure drugs. The data reported herein clearly demonstrate that electrospun PCM/CAF fibers comprise excellent candidates for oral fast-dissolving films, which could be particularly useful for children and patients with swallowing difficulties.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Acetaminophen, analytical standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%