  • Search Results
  • Haemodynamic interactions of medetomidine and the peripheral alpha-2 antagonist MK-467 during step infusions in isoflurane-anaesthetised dogs.
Haemodynamic interactions of medetomidine and the peripheral alpha-2 antagonist MK-467 during step infusions in isoflurane-anaesthetised dogs.
Veterinary journal (London, England : 1997) (2014-09-10)
Johanna Kaartinen, Jérôme R E del Castillo, Kati Salla, Eric Troncy, Marja R Raekallio, Outi M Vainio
ABSTRACT
The haemodynamic interactions of a step infusion with medetomidine (MED) and the peripherally acting alpha-2 antagonist MK-467 (MK) were compared with MED infused alone in isoflurane-anaesthetised dogs. Eight purposely-bred Beagles were used in a randomised crossover study. Anaesthesia was induced with propofol intravenously (IV) and maintained with isoflurane in oxygen. Dogs received 1.25 µg/kg MED as a 1 min loading dose IV, along with a step-down MED infusion at rates of 8.0 µg/kg/h (step 1: 0-20 min), 5.5 µg/kg/h (step 2: 20-40 min) and 4.0 µg/kg/h (step 3: 40-95 min). Five minutes after starting the MED infusion, the dogs received MK-467 in a step-up infusion at rates of 100 µg/kg/h (step 1: 5-35 min), 200 µg/kg/h (step 2: 35-65 min) and 500 µg/kg/h (step 3: 65-95 min). Heart rate (HR), systolic (SAP) and mean arterial (MAP) blood pressures and arteriovenous oxygen content differences (a-vO2 diff) were calculated. Plasma drug concentrations were analysed. Repeated-measures general linear mixed models with Bonferroni correction were used for statistical analyses. MED infusion alone increased SAP maximally by 24.9%, MAP by 34.7% and a-vO2 diff by 222.5%, and reduced HR by 32.3%, but these changes were significantly attenuated by MK-467. Most MED effects returned to baseline during step 2 of MK-467 infusion and step 3 of MED infusion (MED/MK-467 ratio 1:18 to 1:50). Plasma concentrations of MED tended to be lower with the addition of MK-467. The use of step infusions helped to narrow down the therapeutic range for the MED/MK-467 infusion dose ratio during isoflurane anaesthesia in dogs.
Product Description
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
Sigma-Aldrich
Hydrochloric acid solution, SAJ first grade, 9.5-10.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 3% in ethanol
Sigma-Aldrich
Hydrochloric acid, suitable for arsenic determination, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ super special grade, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.1 M
Sigma-Aldrich
Hydrochloric acid, suitable for determination of toxic metals, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M