Merck
  • Antioxidant defense and hepatoprotection by procyanidins from almond (Prunus amygdalus) skins.
Antioxidant defense and hepatoprotection by procyanidins from almond (Prunus amygdalus) skins.
Journal of agricultural and food chemistry (2014-08-15)
Van-Long Truong, Min-Ji Bak, Mira Jun, Ah-Ng Tony Kong, Chi-Tang Ho, Woo-Sik Jeong
ABSTRACT
Procyanidins, polymeric flavan-3-ols, are known to possess antioxidant, antiatherogenic, and anticarcinogenic properties. In the present study, we investigated the role of almond (Prunus amygdalus) skin procyanidins (ASP) in regulating the protein expression of phase II detoxifying and antioxidant enzymes in HepG2 cells and acetaminophen (APAP)-treated hepatotoxic mice. Treatments of ASP significantly induced the expression of phase II enzymes including quinoneoxidoreductase 1, catalase, glutathione peroxidase, and superoxide dismutase in the cells and mice. ASP also potently enhanced the expression of nuclear factor-E2-related factor 2 (Nrf2) and antioxidant response element (ARE)-reporter gene activity in vitro. APAP-induced hepatotoxic markers including AST and ALT in mice were inhibited by ASP administration. However, regulation of upstream kinases by ASP was different between in vitro and in vivo models. Collectively, ASP could induce the activation of Nrf2/ARE-mediated phase II detoxifying/antioxidant enzymes but with differential regulation on upstream kinases between in vitro and in vivo.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%