Merck
  • Luteolin and Gemcitabine Protect Against Pancreatic Cancer in an Orthotopic Mouse Model.
Luteolin and Gemcitabine Protect Against Pancreatic Cancer in an Orthotopic Mouse Model.
Pancreas (2014-09-23)
Jodee L Johnson, Vermont P Dia, Matthew Wallig, Elvira Gonzalez de Mejia
ABSTRACT
This study aimed to evaluate the ability of luteolin (Lut), gemcitabine (Gem), and their combination (Lut + Gem) to prevent the growth of pancreatic tumors in vivo. The antitumor effect of intraperitoneally administered Lut, Gem, and Lut + Gem was evaluated using an orthotopic mouse model for 6 weeks. Tumor growth after injection of human pancreatic cancer cells was assessed by measuring pancreatic tumor mass. The mechanism of action of antitumor effect was assessed by immunohistochemistry and Western blot procedures. Luteolin + Gem significantly lowered (P = 0.048) the pancreatic tumor mass compared with control. Luteolin, Gem, and Lut + Gem significantly reduced the proliferating cell nuclear antigen expression (25%, 37%, and 37%, respectively). Luteolin + Gem treatment led to a significant reduction in the expressions of K-ras (46%, P = 0.0006), GSK-3β (34%, P = 0.014), P(Tyr216)GSK-3β (16%, P = 0.033), P(Ser311)NF-κB p65 (27%, P = 0.036), and bcl-2/bax ratio (68%, P = 0.006) while significantly increasing the expressions of cytochrome c (44%, P = 0.035) and caspase 3 (417%, P = 0.003). Luteolin + Gem promoted apoptotic cell death in pancreatic tumor cells in vivo through inhibition of the K-ras/GSK-3β/NF-κB signaling pathway, leading to a reduction in the Bcl-2/Bax ratio, release of cytochrome c, and activation of caspase 3.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
USP
Gemcitabine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Gemcitabine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Supelco
Luteolin, analytical standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Luteolin, primary reference standard