  • Search Results
  • Synthesis and biological activity of novel mono-indole and mono-benzofuran inhibitors of bacterial transcription initiation complex formation.
Synthesis and biological activity of novel mono-indole and mono-benzofuran inhibitors of bacterial transcription initiation complex formation.
Bioorganic & medicinal chemistry (2015-03-18)
Marcin Mielczarek, Ruth V Thomas, Cong Ma, Hakan Kandemir, Xiao Yang, Mohan Bhadbhade, David StC Black, Renate Griffith, Peter J Lewis, Naresh Kumar
ABSTRACT
Our ongoing research focused on targeting transcription initiation in bacteria has resulted in synthesis of several classes of mono-indole and mono-benzofuran inhibitors that targeted the essential protein-protein interaction between RNA polymerase core and σ(70)/σ(A) factors in bacteria. In this study, the reaction of indole-2-, indole-3-, indole-7- and benzofuran-2-glyoxyloyl chlorides with amines and hydrazines afforded a variety of glyoxyloylamides and glyoxyloylhydrazides. Similarly, condensation of 2- and 7-trichloroacetylindoles with amines and hydrazines delivered amides and hydrazides. The novel molecules were found to inhibit the RNA polymerase-σ(70)/σ(A) interaction as measured by ELISA, and also inhibited the growth of both Gram-positive and Gram-negative bacteria in culture. Structure-activity relationship (SAR) studies of the mono-indole and mono-benzofuran inhibitors suggested that the hydrophilic-hydrophobic balance is an important determinant of biological activity.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, JIS special grade, ≥99.5%
Sigma-Aldrich
Pyridine, suitable for hydroxyl value determination, ≥99.5%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS