  • Search Results
  • Nitric oxide synthase modulates the antihyperalgesic effect of the NMDA receptor antagonist MK-801 on Carrageenan-induced inflammatory pain in rats.
Nitric oxide synthase modulates the antihyperalgesic effect of the NMDA receptor antagonist MK-801 on Carrageenan-induced inflammatory pain in rats.
The Tohoku journal of experimental medicine (2014-12-09)
Dragana P Srebro, Sonja M Vucković, Katarina R Savic Vujovic, Milica S Prostran
ABSTRACT
The N-methyl-D-aspartate (NMDA) receptor, an ionotropic glutamate receptor, may play a significant role in the development and maintenance of an inflammatory pain. Activation of NMDA receptors may cause nitric oxide (NO) release through activation of NO synthase (NOS). MK-801, a noncompetitive NMDA receptor antagonist is commonly used as a neuropharmacological tool. The interaction between MK-801 and NOS in the inflammatory pain has not been evaluated before. We investigated whether MK-801 affects inflammatory pain and whether NOS modulates the effect of MK-801. Carrageenan-induced hyperalgesia was evaluated by measuring the withdrawal response to mechanical stimuli, using an electronic version of the von Frey anesthesiometer in Wistar rats. MK-801 given subcutaneously (0.5-20 μg/kg) or intraplantarly (0.1 and 0.15 μg/paw) significantly reduced mechanical hyperalgesia. Intraplantarly given MK-801 exerted a local antihyperalgesic effect, because when applied to the contralateral side it did not reduce mechanical sensitivity in the ipsilateral side. N-nitro-L-arginine methyl ester hydrochloride (5 and 10 mg/kg), a non-selective NOS inhibitor, significantly reduced the effects of MK-801. N-ω-Propyl-L-arginine hydrochloride (0.5-2 mg/kg), a selective inhibitor of neuronal NOS, increased the antihyperalgesic effect of MK-801, whereas S-methylisothiourea (5-15 μg/kg), a selective inhibitor of inducible NOS, lowered the antihyperalgesic effect of MK-801. Importantly, each NOS inhibitor given alone did not affect carrageenan-induced hyperalgesia. In conclusion, MK-801 is effective against inflammatory pain and its antihyperalgesic effect is modulated in a different ways by NOS, being enhanced by a neuronal NOS inhibitor but reduced by an inducible NOS inhibitor.
Sigma-Aldrich
S-Methylisothiourea hemisulfate salt, 98%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, SAJ first grade, 9.5-10.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 3% in ethanol
Sigma-Aldrich
Hydrochloric acid, suitable for arsenic determination, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ super special grade, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.1 M
Sigma-Aldrich
Hydrochloric acid, suitable for determination of toxic metals, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M