  • Search Results
  • Xanthine oxidase inhibition alleviates the cardiac complications of insulin resistance: effect on low grade inflammation and the angiotensin system.
Xanthine oxidase inhibition alleviates the cardiac complications of insulin resistance: effect on low grade inflammation and the angiotensin system.
Journal of translational medicine (2015-04-19)
Hany M El-Bassossy, Malcolm L Watson
ABSTRACT
We have previously shown that hyperuricemia plays an important role in the vascular complications of insulin resistance (IR). Here we investigated the effect of xanthine oxidase (XO) inhibition on the cardiac complications of IR. IR was induced in rats by a high fructose high fat diet for 12 weeks. Allopurinol, a standard XO inhibitor, was administered in the last 4 weeks before cardiac hemodynamics and electrocardiography, serum glucose, insulin, tumor necrosis factor alpha (TNFα), 8-isoprostane, uric acid, lactate dehydrogenase (LDH) and XO activity were measured. Expression of cardiac angiotensin II (AngII) and angiotensin receptor 1 (AT1) were assessed by immunofluorescence. IR animals had significant hyperuricemia which was inhibited by allopurinol administration. IR was associated with impaired ventricular relaxation (reflected by a decreased diastolic pressure increment and prolonged diastolic duration) and XO inhibition greatly attenuated impaired relaxation. IR was accompanied by cardiac ischemia (reflected by increased QTc and T peak trend intervals) while XO inhibition alleviated the ECG abnormalities. When subjected to isoproterenol-induced ischemia, IR hearts were less resistant (reflected by larger ST height depression and higher LDH level) while XO inhibition alleviated the accompanying ischemia. In addition, XO inhibition prevented the elevation of serum 8-isoprostane and TNFα, and blocked elevated AngII and AT1 receptor expression in the heart tissue of IR animals. However, XO inhibition did not affect the developed hyperinsulinemia or dyslipidemia. XO inhibition alleviates cardiac ischemia and impaired relaxation in IR through the inhibition of low grade inflammation and the angiotensin system.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Supelco
Urethane, analytical standard
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, AR, anhydrous