  • Search Results
  • Miconazole nitrate bearing ultraflexible liposomes for the treatment of fungal infection.
Miconazole nitrate bearing ultraflexible liposomes for the treatment of fungal infection.
Journal of liposome research (2014-02-01)
Jayamanti Pandit, Minakshi Garg, Narendra Kumar Jain
ABSTRACT
Miconazole nitrate is a widely used antifungal agent, but its use in topical formulations is not efficacious because deep seated fungal infections are difficult to treat with conventional topical formulation. Miconazole nitrate loaded ultraflexible liposomes have been prepared and their topical performance has been compared with conventional liposomes containing miconazole nitrate. Various ultraflexible liposomal formulations were prepared and extensively characterized for vesicular shape, size, entrapment efficiency, degree of deformability and in-vitro skin permeation through rat skin. Higher rate of drug transfer across the skin with ultraflexible liposomal formulations of miconazole nitrate suggests that the drug in its lipo-solubilized state might have gained facilitated entry into the tough barrier consisting of subcutaneous. In-vivo study showed better antifungal activity as compared to traditional liposomes and plain drug solution. This was confirmed through fluoroscence microscopy. It is concluded that prepared ultraflexible liposomes can facilitate improved and localized drug action in the skin, thus providing a better option to deal with deep seated skin problems.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Propylene glycol, FCC, FG
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
6-Carboxyfluorescein, ~97% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, AR, anhydrous
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)