Merck
  • LC-MS/MS analysis of pramipexole in mouse plasma and tissues: elimination of lipid matrix effects using weak cation exchange mode based solid-phase extraction.
LC-MS/MS analysis of pramipexole in mouse plasma and tissues: elimination of lipid matrix effects using weak cation exchange mode based solid-phase extraction.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-18)
Weilin Guo, Gao Li, Yanxia Yang, Conglian Yang, Luqin Si, Jiangeng Huang
ABSTRACT
Intranasal delivery is emerging as a promising alternative for oral or intravenous administration of central nervous system (CNS) drugs, such as pramipexole which is widely used for the treatment of Parkinson's disease. To evaluate the effectiveness of intranasal delivery of pramipexole, preclinical pharmacokinetic and tissue distribution studies following intranasal administration need to be investigated. In this paper, we developed and validated a robust and sensitive LC-MS/MS assay without matrix effect for accurate measurements of pramipexole in mouse plasma and tissue samples. Pramipexole and its stable isotope labeled internal standard (d3-pramipexole) were extracted from biological samples by protein precipitation (PPT) coupled with solid phase extraction (SPE) using weak cation exchange SPE cartridges. Matrix effects were studied using post-column infusion and post-extraction addition experiments by direct monitoring of typical phospholipids including glycerophosphocholines (GPChos) and lysoglycerophosphocholines (Lyso-GPChos). Chromatographic separation was achieved on a Welch Ultimate(®) XB-CN column using isocratic elution with a run time of 3.0 min. The assay was linear in the concentration range of 0.05-100 ng/mL and the intra- and inter-day precision and accuracy met the acceptance criteria. Compared with previous reported assays, the current sample preparation approach exhibited significant reduction of matrix effects due to the dramatically decreased levels of residual matrix components such as GPChos and Lyso-GPChos. This method has been successfully applied to pharmacokinetic and tissue distribution studies of pramipexole in mice following a single intravenous or intranasal dose of 50 μg/kg.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Dichloromethane, HPLC grade
Sigma-Aldrich
Dichloromethane, JIS 5000, for residue analysis
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Dichloromethane, JIS 300, for residue analysis
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Dichloromethane, analytical standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)