  • Search Results
  • Validated stability indicating RP-HPLC for quantitation of nitazoxanide in presence of its alkaline degradation products and their characterization by HPLC-tandem mass spectrometry.
Validated stability indicating RP-HPLC for quantitation of nitazoxanide in presence of its alkaline degradation products and their characterization by HPLC-tandem mass spectrometry.
Journal of chromatographic science (2013-11-20)
Maha Hegazy, Amira Kessiba, Ahmed Emad El Gindy, Mohamed Abdelkawy
ABSTRACT
A simple and sensitive stability indicating HPLC method was developed and validated for quantitative determination of Nitazoxanide (NTZ), a new antiprotozoal drug, in presence of degradation products generated under forced alkaline hydrolysis. Chromatographic separation was achieved on Inertsil C8-3 column (150 × 4.6 mm i.d.) using a mobile phase composed of acetonitrile: 50 mM ammonium acetate buffer (50:50, v/v, pH 5.0 adjusted with acetic acid) at a flow rate of 1 mL/min. Quantification was achieved with UV detection at 298 nm based on relative peak area. The method was linear over the concentration range of 0.8-50 µg/mL (r = 0.9999) with a limit of detection and quantification 0.0410 and 0.1242 µg/mL, respectively. The developed method has the requisite accuracy, selectivity, sensitivity and precision to assay NTZ in presence of its degradation products either in bulk powder or in pharmaceutical formulations. The degradation products were then identified by HPLC-MS/MS analysis using an electrospray ionization source and an ion trap analyzer.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS