  • Search Results
  • Floating lipid beads for the improvement of bioavailability of poorly soluble basic drugs: in-vitro optimization and in-vivo performance in humans.
Floating lipid beads for the improvement of bioavailability of poorly soluble basic drugs: in-vitro optimization and in-vivo performance in humans.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-03)
Samar M Abouelatta, Ahmed A Aboelwafa, Rawia M Khalil, Omaima N ElGazayerly
ABSTRACT
The challenge in developing oral drug delivery systems of poorly soluble basic drugs is primarily due to their pH dependent solubility. Cinnarizine (CNZ), a model for a poorly soluble basic drug, has pH dependent solubility; where it dissolves readily at low pH in the stomach and exhibits a very low solubility at pH values greater than 4. It is also characterized by a short half life of 3-6h, which requires frequent daily administration resulting in poor patient compliance. In an attempt to solve these problems, extended release floating lipid beads were formulated. A 2(4) full factorial design was utilized for optimization of the effects of various independent variables; lipid:drug ratio, % Pluronic F-127, % Sterotex, and Gelucire 43/01:Gelucire 50/13 ratio, on the loading efficiency and release of CNZ from the lipid beads. In-vivo pharmacokinetic study of the optimized CNZ-lipid beads compared to Stugeron® (reference standard) was performed in healthy human volunteers. A promising approach for enhancing the bioavailability of the poorly soluble basic drug, CNZ, utilizing novel and simple floating lipid beads was successfully developed. Zero order release profile of CNZ was achieved for 12h. Mean AUC0-24 and AUC0-∞ of the optimized CNZ-loaded lipid beads were 4.23 and 6.04 times that of Stugeron® tablets respectively.
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, AR, ≥99.5%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether