  • Search Results
  • FBXL5-mediated degradation of single-stranded DNA-binding protein hSSB1 controls DNA damage response.
FBXL5-mediated degradation of single-stranded DNA-binding protein hSSB1 controls DNA damage response.
Nucleic acids research (2014-09-25)
Zhi-Wei Chen, Bin Liu, Nai-Wang Tang, Yun-Hua Xu, Xiang-Yun Ye, Zi-Ming Li, Xiao-Min Niu, Sheng-Ping Shen, Shun Lu, Ling Xu
ABSTRACT
Human single-strand (ss) DNA binding proteins 1 (hSSB1) has been shown to participate in DNA damage response and maintenance of genome stability by regulating the initiation of ATM-dependent signaling. ATM phosphorylates hSSB1 and prevents hSSB1 from ubiquitin-proteasome-mediated degradation. However, the E3 ligase that targets hSSB1 for destruction is still unknown. Here, we report that hSSB1 is the bona fide substrate for an Fbxl5-containing SCF (Skp1-Cul1-F box) E3 ligase. Fbxl5 interacts with and targets hSSB1 for ubiquitination and degradation, which could be prevented by ATM-mediated hSSB1 T117 phosphorylation. Furthermore, cells overexpression of Fbxl5 abrogated the cellular response to DSBs, including activation of ATM and phosphorylation of ATM targets and exhibited increased radiosensitivity, chemosensitivity and defective checkpoint activation after genotoxic stress stimuli. Moreover, the protein levels of hSSB1 and Fbxl5 showed an inverse correlation in lung cancer cells lines and clinical lung cancer samples. Therefore, Fbxl5 may negatively modulate hSSB1 to regulate DNA damage response, implicating Fbxl5 as a novel, promising therapeutic target for lung cancers.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Etoposide, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Caffeine, European Pharmacopoeia (EP) Reference Standard