  • Search Results
  • Adiponectin stimulates Rho-mediated actin cytoskeleton remodeling and glucose uptake via APPL1 in primary cardiomyocytes.
Adiponectin stimulates Rho-mediated actin cytoskeleton remodeling and glucose uptake via APPL1 in primary cardiomyocytes.
Metabolism: clinical and experimental (2014-08-12)
Rengasamy Palanivel, Riya Ganguly, Subat Turdi, Aimin Xu, Gary Sweeney
ABSTRACT
Adiponectin is known to confer its cardioprotective effects in obesity and type 2 diabetes, mainly by regulating glucose and fatty acid metabolism in cardiomyocytes. Dynamic actin cytoskeleton remodeling is involved in regulation of multiple biological functions, including glucose uptake. Here we investigated in neonatal cardiomyocytes whether adiponectin induced actin cytoskeleton remodeling and if this played a role in adiponectin-stimulated glucose uptake. Primary cardiomyocytes were treated with full-length and globular adiponectin (fAd and gAd, respectively). Both fAd and gAd increased RhoA activity, phosphorylation of the Rho/ROCK signaling target cofilin and actin polymerization to form filamentous actin as determined by rhodamine-phallodin immunofluorescence and quantitative analysis of filamentous to globular actin ratio. Scanning electron microscopy also demonstrated structural remodeling. Adiponectin stimulated glucose uptake, was significantly abrogated in the presence of inhibitors of actin cytoskeleton remodeling (cytochalasin D) and Rho/ROCK signaling (C3 transferase, Y27632). We showed that adiponectin increased colocalization of actin and APPL1 and that actin remodeling, phosphorylation of AMPK, p38MAPK and cofilin, glucose uptake and oxidation were all attenuated after siRNA-mediated knockdown of APPL1. We show that adiponectin mediates Rho/ROCK-dependent actin cytoskeleton remodeling to increase glucose uptake and metabolism via APPL1 signaling.
Sigma-Aldrich
Glucose stock solution, 1000 mg/dL
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material