  • Search Results
  • Type AII lantibiotic bovicin HJ50 with a rare disulfide bond: structure, structure-activity relationships and mode of action.
Type AII lantibiotic bovicin HJ50 with a rare disulfide bond: structure, structure-activity relationships and mode of action.
The Biochemical journal (2014-05-13)
Jie Zhang, Yingang Feng, Kunling Teng, Yuheng Lin, Yong Gao, Jinfeng Wang, Jin Zhong
ABSTRACT
Lantibiotics are ribosomally synthesized antimicrobial peptides containing unusual amino acids. As promising alternatives to conventional antibiotics, they have a high potential for alleviating the problem of emergent antibiotic resistance, with possible applications in many industries that have antibacterial demand. Bovicin HJ50 is a type AII lantibiotic, the largest group of lantibiotics, comprising a linear N-terminal region and a globular C-terminal region. Interestingly, bovicin H50 has a disulfide bond that is rare in this group. Owing to limited information about the spatial structures of type AII lantibiotics, the functional regions of this type and the role of the disulfide bond are still unknown. In the present study, we resolved the solution structure of bovicin HJ50 using NMR spectroscopy. This is the first spatial structure of a type AII lantibiotic. Bovicin HJ50 exhibited high flexibility in aqueous solution, whereas varied rigidities were observed in the different rings with the conserved ring A being the most rigid. The charged residues Lys¹¹, Asp¹² and Lys³⁰, as well as the essential disulfide bond were critical for antimicrobial activity. Importantly, bovicin HJ50 showed not only peptidoglycan precursor lipid II-binding ability, but also pore-forming activity, which is significantly different from other bacteriostatic type AII lantibiotics, suggesting a novel antimicrobial mechanism.
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Cystine
Supelco
L-Cystine, certified reference material, TraceCERT®
SAFC
L-Cystine
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cystine, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Aspartic acid, SAJ special grade, ≥99.0%
Supelco
L-Aspartic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard