Merck
  • Role of tautomerism of 2-azaadenine and 2-azahypoxanthine in substrate recognition by xanthine oxidase.
Role of tautomerism of 2-azaadenine and 2-azahypoxanthine in substrate recognition by xanthine oxidase.
Journal of computer-aided molecular design (1997-03-01)
B Hernández, M Orozco, F J Luque
ABSTRACT
The tautomerism of 2-azaadenine and 2-hypoxanthine has been examined in the gas phase and in aqueous solution. The tautomerism in the gas phase has been studied by means of semiempirical and ab initio quantum-mechanical computations, as well as density-functional calculations. The influence of the aqueous solvent on the relative stability between tautomers has been estimated from self-consistent reaction field calculations performed with different high-level continuum models. The results provide a detailed picture of the tautomeric preference for these purine bases. The importance of tautomerism in the substrate recognition by xanthine oxidase is discussed. Finally, the rate of oxidation of 2-azaadenine and 2-hypoxanthine by xanthine oxidase is discussed in terms of the recognition model at the enzyme active site.