  • Search Results
  • Compatibility of amiodarone hydrochloride with vasopressin during simulated Y-site administration.
Compatibility of amiodarone hydrochloride with vasopressin during simulated Y-site administration.
International journal of pharmaceutical compounding (2014-01-28)
Corinne C Ramaley, Feng Li, Chengan Du
ABSTRACT
The objective of this study was to determine the compatibility of amiodarone hydrochloride with vasopressin during simulated Y-site administration. Amiodarone hydrochloride is compatible with vasopressin under simulated-use conditions when administered according to the Advanced Cardiac Life Support Guidelines. A simulated-use approach is a more appropriate way to evaluate compatibility of drug solutions and to assure safe and adequate drug delivery to the patient, particularly when the drug being evaluated is well-documented to have physical or chemical incompatibility concerns with various drugs. When amiodarone hydrochloride (50 mg/mL) was diluted to 6 mg/mL with 5% dextrose injection, then mixed with an equal volume of vasopressin (0.2 units/mL in normal saline) in a glass test tube, no visual incompatibility was observed for up to 24 hours. The purpose of our study was to assess compatibility of the two drugs under simulated-use conditions and to measure the recovery of amiodarone by high-performance liquid chromatography after sequential administration of vasopressin and amiodarone hydrochloride according to the Advanced Cardiac Life Support Guidelines.
Sigma-Aldrich
Glucose stock solution, 1000 mg/dL
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, AR, anhydrous