Merck
  • Characterization of recombinant human adipocyte-derived leucine aminopeptidase expressed in Chinese hamster ovary cells.
Characterization of recombinant human adipocyte-derived leucine aminopeptidase expressed in Chinese hamster ovary cells.
Journal of biochemistry (2000-11-01)
A Hattori, K Kitatani, H Matsumoto, S Miyazawa, T Rogi, N Tsuruoka, S Mizutani, Y Natori, M Tsujimoto
ABSTRACT
Adipocyte-derived leucine aminopeptidase (A-LAP) is a recently identified novel member of the M1 family of zinc-metallopeptidases. Transfection of the A-LAP cDNA into COS-7 cells resulted in the secretion of the enzyme. In this study, recombinant A-LAP was expressed in Chinese hamster ovary cells, purified to homogeneity and its enzymatic properties were characterized. The purified enzyme was active towards a synthetic substrate, L-leucyl-p-nitroanilide, yielding a V(max) of 3.55 micromol/min/mg and a K(m) of 1.28 mM, and was shown to be a monomeric protein with molecular mass of 120 kDa in solution. By monitoring the sequential N-terminal amino acid liberation, it was found that the enzyme hydrolyzes a variety of bioactive peptides, including angiotensin II and kallidin. Immunohistochemical analysis indicated that the enzyme is expressed in the cortex of the human kidney, where tissue kallikrein is localized. Taken together, these results indicate that A-LAP possesses a broad substrate specificity towards naturally occurring peptide hormones and suggest that it plays a role in the regulation of blood pressure through the inactivation of angiotensin II and/or the generation of bradykinin in the kidney.