  • Search Results
  • [Analysis of methionine metabolism studied by the gas chromatographic determination of 3-methylthiopropionate in urine and its clinical application].
[Analysis of methionine metabolism studied by the gas chromatographic determination of 3-methylthiopropionate in urine and its clinical application].
[Hokkaido igaku zasshi] The Hokkaido journal of medical science (1985-03-01)
Y Yoshida
ABSTRACT
Methionine is a sulfur-containing essential amino acid and there are optic isomers of L- and D-type. It has been known that there are two metabolic pathways in methionine metabolism-the transsulfuration and the transamination. The purposes of the present investigation are to clarify the existence of transaminative pathway in human by the quantitative analysis of 3-methylthiopropionate (3-MTP), one of the metabolites of methionine, and to study its clinical significance in patients with liver cirrhosis. 3-MTP in urine was analysed by gas chromatograph equipped with the flame photometric detector (FPD) which has a highly specific sensitivity to the sulfur compounds. Fasting levels of 3-MTP concentration in urine in healthy subjects (n = 20) and patients with liver cirrhosis (n = 21) were 39.1 +/- 9.7 ng/mg. Cr. (Mean +/- SE) and 103.6 +/- 24.2 ng/mg. Cr., respectively. 3-MTP concentration in urine in cirrhotic patients was significantly higher than that in healthy subjects (p less than 0.05). In some cases, 3-MTP concentration in urine was measured every hour for 3 to 6 hours after the oral loading of 2 g of D- or L-methionine. In healthy subjects, 3-MTP concentration in urine increased remarkably at 1 hour period after oral loading of 2 g of D-methionine, and subsequently its concentration showed a tendency to decrease gradually. However, there were no such changes after oral loading of 2 g of L-methionine. When 2 g of D-methionine was loaded orally, decreasing curves of 3-MTP concentration in urine were different between healthy subjects and patients with liver cirrhosis, and half-disappearance time of 3-MTP concentration in urine was remarkably prolonged in cirrhotic patients. These findings seem to indicate that 3-MTP is one of the metabolites of methionine (especially of D-methionine) and the transaminative pathway exists in human. It was also suggested that there was the impairment of the transaminative pathway of methionine metabolism in patients with liver cirrhosis. Pharmacokinetics of 3-MTP in urine seems to contribute to the clinicopathological investigation of the liver cirrhosis.
Sigma-Aldrich
Propylene glycol, FCC, FG
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
2,3-Butanediol, 98%
SAFC
L-Aspartic acid
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, SAJ special grade, ≥99.0%
Supelco
L-Aspartic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Aspartic acid, ≥98%, FG
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%