  • Search Results
  • Glycolamide esters as biolabile prodrugs of carboxylic acid agents: synthesis, stability, bioconversion, and physicochemical properties.
Glycolamide esters as biolabile prodrugs of carboxylic acid agents: synthesis, stability, bioconversion, and physicochemical properties.
Journal of pharmaceutical sciences (1988-04-01)
N M Nielsen, H Bundgaard
ABSTRACT
Benzoic acid esters of various substituted 2-hydroxyacetamides (glycolamides) were found to be hydrolyzed extremely rapidly in human plasma solutions, the half-lives of hydrolysis being less than 5 s in 50% plasma solutions for some N,N-disubstituted glycolamide esters. The rapid rate of hydrolysis could be largely attributed to cholinesterase (also called pseudocholinesterase) present in plasma. From a study of a variety of substituted glycolamide esters and structurally related esters, the most prominent structural requirement needed for a rapid rate of hydrolysis was found to be the glycolamide ester structure combined with the presence of two substituents on the amide nitrogen atom. A structural similarity of such esters with benzoylcholine, a good substrate for cholinesterase, was put forward. Esters of N,N-disubstituted glycolamides are suggested to be a useful biolabile prodrug type for several carboxylic acid agents. The esters combine a high susceptibility to undergo enzymatic hydrolysis in plasma with a high stability in aqueous solution. Furthermore, as demonstrated with the benzoic acid model esters, it is feasible to obtain ester derivatives with almost any desired water solubility or lipophilicity with retainment of marked lability to enzymatic hydrolysis.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Supelco
Benzoic acid, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®