  • Search Results
  • Percutaneous absorption of benzoic acid across human skin. I. In vitro experiments and mathematical modeling.
Percutaneous absorption of benzoic acid across human skin. I. In vitro experiments and mathematical modeling.
Pharmaceutical research (1990-03-01)
G E Parry, A L Bunge, G D Silcox, L K Pershing, D W Pershing
ABSTRACT
The percutaneous absorption of benzoic acid across human skin in vitro was experimentally and mathematically modeled. Skin partition coefficients were measured over a range of benzoic acid concentrations in both saline and distilled water. The permeation of benzoic acid was measured across isolated stratum corneum, stratum corneum and epidermis, and split-thickness skin. These experiments demonstrated that the stratum corneum was the rate-limiting barrier and that the flux is proportional to the concentration of the undissociated species. The permeation data were analyzed with a comprehensive non-steady-state mathematical model of diffusion across skin. Two adjustable parameters, the effective skin thickness and diffusivity, were fit to the permeation data by nonlinear regression.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Supelco
Benzoic acid, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®
Sigma-Aldrich
Benzoic acid, AR, ≥99%
Sigma-Aldrich
Benzoic acid, LR, ≥99%