  • Search Results
  • Hepatic veno-occlusive disease due to tacrolimus in a single-lung transplant patient.
Hepatic veno-occlusive disease due to tacrolimus in a single-lung transplant patient.
The European respiratory journal (2006-05-19)
S Shah, M Budev, H Blazey, K Fairbanks, A Mehta
ABSTRACT
Hepatic veno-occlusive disease is defined as nonthrombotic fibrous obliterative endophlebitis of small centrilobular hepatic venules. Clinically, patients present with elevated liver enzymes and a triad of jaundice, hepatomegaly and ascites. Although reported as a complication of other solid organ and stem cell transplantation, there have been no reported cases to date of veno-occlusive disease following lung transplantation. The present authors report a case of veno-occlusive disease following single-lung transplantation in a patient on a triple-drug immunosuppressive regimen composed of tacrolimus, mycophenolate mofetil and prednisone. The diagnosis was established by transjugular liver biopsy and by discontinuing tacrolimus; there was clinical regression of symptoms and serological return to baseline.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard