  • Search Results
  • Chlorogenic acid, quercetin-3-rutinoside and black tea phenols are extensively metabolized in humans.
Chlorogenic acid, quercetin-3-rutinoside and black tea phenols are extensively metabolized in humans.
The Journal of nutrition (2003-05-29)
Margreet R Olthof, Peter C H Hollman, Michel N C P Buijsman, Johan M M van Amelsvoort, Martijn B Katan
ABSTRACT
Dietary phenols are antioxidants, and their consumption might contribute to the prevention of cardiovascular disease. Coffee and tea are major dietary sources of phenols. Dietary phenols are metabolized extensively in the body. Lack of quantitative data on their metabolites hinders a proper evaluation of the potential biological effects of dietary phenols in vivo. The aim of this study was to identify and quantify the phenolic acid metabolites of chlorogenic acid (major phenol in coffee), quercetin-3-rutinoside (major flavonol in tea) and black tea phenols in humans, and determine the site of metabolism. Healthy humans (n = 20) with an intact colon participated in a dietary controlled crossover study, and we identified and quantified approximately 60 potential phenolic acid metabolites in urine. Half of the ingested chlorogenic acid and 43% of the tea phenols were metabolized to hippuric acid. Quercetin-3-rutinoside was metabolized mainly to phenylacetic acids, i.e., 3-hydroxyphenylacetic acid (36%), 3-methoxy-4-hydroxyphenylacetic acid (8%) and 3,4-dihydroxyphenylacetic acid (5%). In contrast, in seven humans without a colon, we found only traces of phenolic acid metabolites in urine after they had ingested chlorogenic acid and quercetin-3-rutinoside. This implies that the colonic microflora convert most of these dietary phenols into metabolites that then reach the circulation. Metabolites of dietary phenols have lower antioxidant activity than their parent compounds; therefore, the contribution of dietary phenols to antioxidant activity in vivo might be lower than expected from in vitro tests.
Sigma-Aldrich
2-Hydroxyphenylacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Supelco
Vanillic acid, certified reference material, TraceCERT®
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
4-Hydroxybenzoic acid, ≥99%, FG
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%