  • Search Results
  • [Antidepressant active constituents in the roots of Morinda officinalis How].
[Antidepressant active constituents in the roots of Morinda officinalis How].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1995-01-01)
C Cui, M Yang, Z Yao, B Cao, Z Luo, Y Xu, Y Chen
ABSTRACT
Five compounds having antidepressant activities have been isolated from the roots of Morinda officinalis, a Chinese traditional Yang-tonic drug. These compounds were identified as succinic acid (1), nystose (2), 1F-fructofuranosylnystose (3), inulin-type hexasaccharide (4) and heptasaccharide (5) by chemical and spectroscopic methods. All of the compounds are isolated from the species of genus Morinda for the first time.
Product Description
Supelco
Nystose, analytical standard