  • Search Results
  • Changes in taste neurons support the emergence of an adaptive behavior in cockroaches.
Changes in taste neurons support the emergence of an adaptive behavior in cockroaches.
Science (New York, N.Y.) (2013-05-25)
Ayako Wada-Katsumata, Jules Silverman, Coby Schal
ABSTRACT
In response to the anthropogenic assault of toxic baits, populations of the German cockroach have rapidly evolved an adaptive behavioral aversion to glucose (a phagostimulant component of baits). We hypothesized that changes in the peripheral gustatory system are responsible for glucose aversion. In both wild-type and glucose-averse (GA) cockroaches, D-fructose and D-glucose stimulated sugar-gustatory receptor neurons (GRNs), whereas the deterrent caffeine stimulated bitter-GRNs. In contrast, in GA cockroaches, D-glucose also stimulated bitter-GRNs and suppressed the responses of sugar-GRNs. Thus, D-glucose is processed as both a phagostimulant and deterrent in GA cockroaches, and this newly acquired peripheral taste sensitivity underlies glucose aversion in multiple GA populations. The rapid emergence of this highly adaptive behavior underscores the plasticity of the sensory system to adapt to rapid environmental change.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Caffeine, European Pharmacopoeia (EP) Reference Standard