Merck
  • Internal standard application to dried blood spots by spraying: investigation of the internal standard distribution.
Internal standard application to dried blood spots by spraying: investigation of the internal standard distribution.
Bioanalysis (2013-03-15)
Dieter Zimmer, Stephan Hassler, Beatrice Betschart, Stephan Sack, Christoph Fankhauser, Matthias Loppacher
ABSTRACT
The scientifically and logistically best way of application of the internal standard (IS) in the analysis of dried blood spots (DBS) analysis is still a matter of debate and investigation. Most commonly the IS is added in the solvent used for extraction of the discs punched from DBS. In this case, the recovery of the non-extracted IS is complete while the recovery of the analyte extracted from DBS is different from the IS. An alternative way for addition of the IS was investigated. A homogeneous distribution and absorption of the test compound across the spots was demonstrated by spraying a solution of a radiolabeled test compound (mimicking an IS solution) onto DBS. This spray-on technique is convenient and easily automatable. Spraying of the solution was rapid, precise and reproducible, and therefore seems to be suitable for routine analysis of DBS by offline and online extraction.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Supelco
Caffeine, certified reference material, TraceCERT®
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®