  • Search Results
  • The improvement of the anticancer effect of a novel compound benzoic acid, 2-hydroxy-, 2-D-ribofuranosylhydrazide (BHR) loaded in solid lipid nanoparticles.
The improvement of the anticancer effect of a novel compound benzoic acid, 2-hydroxy-, 2-D-ribofuranosylhydrazide (BHR) loaded in solid lipid nanoparticles.
AAPS PharmSciTech (2012-10-12)
Mei Wang, Lili Qin, Kun Li, Rongrong Zhu, Wenrui Wang, Shilong Wang
ABSTRACT
A novel drug delivery system consisting of benzoic acid, 2-hydroxy-, 2-D-ribofuranosylhydrazide (BHR)-loaded solid lipid nanoparticles (BHR-SLNs) was prepared using the emulsification-evaporation technique. The mean particle size of the BHR-SLNs measured by photon correlation spectroscopy was about 75 nm. BHR-SLN morphology was assessed by transmission electron microscopy and atomic force microscopy. The drug entrapment efficiency was 70.2%, as determined via Sephadex gel chromatography and high-performance liquid chromatography. Drug release assessment in vitro showed that BHR was gradually released from SLNs in a time-dependent manner. Furthermore, treatment of 293T and Hela cells with BHR-SLNs demonstrated that BHR-SLNs were less toxic to normal cells while more effective in antitumor potency compared with the BHR drug alone. The results imply that BHR-SLNs could be considered as a promising antitumor drug system for a range of new therapeutic applications.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Supelco
Benzoic acid, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®