- [Cerebral air embolism complicating talc pleurodesis: case report].
[Cerebral air embolism complicating talc pleurodesis: case report].
Annales francaises d'anesthesie et de reanimation (2012-05-29)
L Gamerre, M Gazon, B Delafosse, J-P Viale
PMID22632995
ABSTRACT
Cerebral air embolism is a severe complication which can occur during many invasive medical procedures. Its prognosis depends on early diagnosis and hyperbaric oxygen therapy. We report a case of cerebral air embolism which occurred during a talc pleurodesis within a chest tube, in chest postoperative period. After early and appropriate treatment, we observed a significant functional recovery.
MATERIALS