  • Search Results
  • Industrial production of fructooligosaccharides by immobilized cells of Aureobasidium pullulans in a packed bed reactor.
Industrial production of fructooligosaccharides by immobilized cells of Aureobasidium pullulans in a packed bed reactor.
Biotechnology letters (2011-04-12)
K H Jung, S H Bang, T K Oh, H J Park
ABSTRACT
Continuous production of fructooligosaccharides (FOS) by Aureobasidium pullulans immobilized on calcium alginate beads with a packed bed was investigated at a plant scale reactor. Optimum conditions were with 770 g sucrose/l, being fed at 200 l/h at 50°C which gave a productivity of 180 g FOS/l h. Initial activity was maintained for more than 100 days. The reactor was successfully scaled up to a production scale of 1.2 m(3).
Product Description
Supelco
Nystose, analytical standard
Supelco
1-Kestose, analytical standard