Merck
An investigation of the ionophoric characteristics of destruxin A.
Archives of biochemistry and biophysics (2002-08-15)
Maria Hinaje, Martyn Ford, Lee Banting, Steve Arkle, Bhupinder Khambay
ABSTRACT
Destruxin A, a cyclohexadepsipeptide related to the enniatins and beauvericin, exhibits ionophoric properties. Calcium ion mobilization across liposomal membrane barriers, for example, has been demonstrated using the calcium ion-sensitive dyes Arsenazo III and Fura-2. Initial molecular mechanics/molecular dynamics calculations indicate the potential for destruxin A to form a coordination complex with calcium in which the divalent cation is bound at the center of a sandwich formed by two molecules of destruxin A. This novel calcium ion binding may help explain the diverse biological effects exhibited by the destruxins.
Sigma-Aldrich
Destruxin A from Metarrhizium anisopliae, ≥98% (HPLC)