Merck
  • Effect of liquiritigenin, a flavanone existed from Radix glycyrrhizae on pro-apoptotic in SMMC-7721 cells.
Effect of liquiritigenin, a flavanone existed from Radix glycyrrhizae on pro-apoptotic in SMMC-7721 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2009-01-20)
Shi-ping Zhang, Yi-jing Zhou, Yan Liu, Yun-qing Cai
ABSTRACT
Liquiritigenin is a flavanone existed in Radix glycyrrhizae. The objective of this study is to explore the effects of liquiritigenin on SMMC-7721 cells and its possible mechanism. The viability of liquiritigenin treat cells was decreased in a dose-dependent manner assayed by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenylterazolium bromide (MTT), and apoptotic morphological changes also be observed, such as chromatin condensation and nuclear fragmentation. Assessment of apoptotic cells by flow cytometry indicated that cells fell into apoptosis after 0.4mM liquiritigenin treatment. In addition, a concomitant time-dependent increase in caspase-3 activity was also observed. The level of p53 protein increased and Bcl-2 protein decreased time-dependently. Further studies found the induction of apoptosis by liquiritigenin was accompanied with the production of reactive oxygen species (ROS), disruption of mitochondrial membrane potential and depletion of antioxidant enzymes. The significant ROS generation was firstly found at 3h and being time-dependent until 9h. A time-dependent decrease in membrane potential occurred, and significant loss appeared at 9h and 12h. Furthermore, pretreatment of N-acetyl-cysteine (NAC), ROS production and apoptosis induced by liquiritigenin were both suppressed. In sum, this paper indicated the cytotoxicity of liquiritigenin on SMMC-7721 cells may via effect on generation of ROS, later lead to cell apoptosis.
Sigma-Aldrich
Liquiritigenin, ≥97.0% (HPLC)