  • Search Results
  • Efficient synthesis and biological evaluation of epiceanothic acid and related compounds.
Efficient synthesis and biological evaluation of epiceanothic acid and related compounds.
Bioorganic & medicinal chemistry letters (2010-12-03)
Pu Zhang, Li Xu, Keduo Qian, Jun Liu, Luyong Zhang, Kuo-Hsiung Lee, Hongbin Sun
ABSTRACT
Epiceanothic acid (1) is a naturally occurring, but very rare pentacyclic triterpene with a unique pentacyclic triterpene (PT) structure. An efficient synthesis of 1 starting from betulin (3) has been accomplished in 12-steps with a total yield of 10% in our study. Compound 1 and selected synthetic intermediates were further evaluated as anti-HIV-1 agents, inhibitors of glycogen phosphorylase (GP), and cytotoxic agents. Compound 1 exhibited moderate HIV-1 inhibition. Most importantly, compound 5, with an opened A-ring, showed significant GP inhibitory activity with an IC(50) of 0.21 μM, suggesting a potential for development as an anti-diabetic agent. On the other hand, compound 12, with a closed A-ring, showed potent cytotoxicity against A549 and MCF-7 human tumor cell lines, with IC(50) values of 0.89 and 0.33 μM, respectively. These results suggest that the A-ring of PTs is an important pharmacophore that could be modified to involve different biological activities.
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Betulin, ≥98%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Supelco
Melting point standard 235-237°C, analytical standard