Merck
  • Cdh1 inhibits WWP2-mediated ubiquitination of PTEN to suppress tumorigenesis in an APC-independent manner.
Cdh1 inhibits WWP2-mediated ubiquitination of PTEN to suppress tumorigenesis in an APC-independent manner.
Cell discovery (2016-07-28)
Jia Liu, Lixin Wan, Jing Liu, Zhu Yuan, Jinfang Zhang, Jianfeng Guo, Marcos Malumbres, Jiankang Liu, Weiguo Zou, Wenyi Wei
ABSTRACT
Anaphase-promoting complex/cyclosome/Cdh1 is a multi-subunit ubiquitin E3 ligase that drives M to G1 cell cycle progression through primarily earmarking various substrates for ubiquitination and subsequent degradation by the 26S proteasome. Notably, emerging evidence suggested that Cdh1 could also function in various cellular processes independent of anaphase-promoting complex/cyclosome. To this end, we recently identified an anaphase-promoting complex/cyclosome-independent function of Cdh1 in modulating osteoblast differentiation through activating Smurf1, one of the NEDD4 family of HECT domain-containing E3 ligases. However, it remains largely unknown whether Cdh1 could exert its tumor suppressor role through similarly modulating the E3 ligase activities of other NEDD4 family members, most of which have characterized important roles in tumorigenesis. Here we report that in various tumor cells, Cdh1, conversely, suppresses the E3 ligase activity of WWP2, another NEDD4 family protein, in an anaphase-promoting complex/cyclosome-independent manner. As such, loss of Cdh1 activates WWP2, leading to reduced abundance of WWP2 substrates including PTEN, which subsequently activates PI3K/Akt oncogenic signaling to facilitate tumorigenesis. This study expands the non-anaphase-promoting complex/cyclosome function of Cdh1 in regulating the NEDD4 family E3 ligases, and further suggested that enhancing Cdh1 to inhibit the E3 ligase activity of WWP2 could be a promising strategy for treating human cancers.
Sigma-Aldrich
Caspase Inhibitor, Negative Control, The Caspase Inhibitor, Negative Control, also referenced under CAS 105637-38-5, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Anti-Cdh1 (Ab-2) Mouse mAb (DH01), liquid, clone DH01, Calbiochem®