  • Search Results
  • Rynaxypyr: a new insecticidal anthranilic diamide that acts as a potent and selective ryanodine receptor activator.
Rynaxypyr: a new insecticidal anthranilic diamide that acts as a potent and selective ryanodine receptor activator.
Bioorganic & medicinal chemistry letters (2007-09-22)
George P Lahm, Thomas M Stevenson, Thomas P Selby, John H Freudenberger, Daniel Cordova, Lindsey Flexner, Cheryl A Bellin, Christine M Dubas, Ben K Smith, Kenneth A Hughes, J Gary Hollingshaus, Christopher E Clark, Eric A Benner
ABSTRACT
Rynaxypyr is a highly potent and selective activator of insect ryanodine receptors with exceptional activity on a broad range of Lepidoptera. A strong correlation between insecticidal activity and ryanodine receptor activation is observed along with selective activity against insect over mammalian receptors. The synthesis and biological results are presented.
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard