Merck
  • A novel phospho-modulatory mechanism contributes to the calcium-dependent regulation of T-type Ca2+ channels.
A novel phospho-modulatory mechanism contributes to the calcium-dependent regulation of T-type Ca2+ channels.
Scientific reports (2019-11-02)
Jean Chemin, Tamara Timic Stamenic, Magalie Cazade, Jodie Llinares, Iulia Blesneac, Slobodan M Todorovic, Philippe Lory
ABSTRACT
Cav3 / T-type Ca2+ channels are dynamically regulated by intracellular Ca2+ ions, which inhibit Cav3 availability. Here, we demonstrate that this inhibition becomes irreversible in the presence of non-hydrolysable ATP analogs, resulting in a strong hyperpolarizing shift in the steady-state inactivation of the residual Cav3 current. Importantly, the effect of these ATP analogs was prevented in the presence of intracellular BAPTA. Additional findings obtained using intracellular dialysis of inorganic phosphate and alkaline phosphatase or NaN3 treatment further support the involvement of a phosphorylation mechanism. Contrasting with Cav1 and Cav2 Ca2+ channels, the Ca2+-dependent modulation of Cav3 channels appears to be independent of calmodulin, calcineurin and endocytic pathways. Similar findings were obtained for the native T-type Ca2+ current recorded in rat thalamic neurons of the central medial nucleus. Overall, our data reveal a new Ca2+ sensitive phosphorylation-dependent mechanism regulating Cav3 channels, with potentially important physiological implications for the multiple cell functions controlled by T-type Ca2+ channels.
Sigma-Aldrich
CGP 57380, ≥98% (HPLC)