Merck
  • The NRON complex controls circadian clock function through regulated PER and CRY nuclear translocation.
The NRON complex controls circadian clock function through regulated PER and CRY nuclear translocation.
Scientific reports (2019-08-17)
Yool Lee, Yang Shen, Lauren J Francey, Chidambaram Ramanathan, Amita Sehgal, Andrew C Liu, John B Hogenesch
ABSTRACT
Post-translational regulation plays a central role in the circadian clock mechanism. However, nucleocytoplasmic translocation of core clock proteins, a key step in circadian timekeeping, is not fully understood. Earlier we found that the NRON scaffolding complex regulates nuclear translocation of NFAT and its signaling. Here, we show that components of the NRON complex also regulate the circadian clock. In peripheral cell clock models, genetic perturbation of the NRON complex affects PER and CRY protein nuclear translocation, dampens amplitude, and alters period length. Further, we show small molecules targeting the NFAT pathway alter nuclear translocation of PER and CRY proteins and impact circadian rhythms in peripheral cells and tissue explants of the master clock in the suprachiasmatic nucleus. Taken together, these studies highlight a key role for the NRON complex in regulating PER/CRY subcellular localization and circadian timekeeping.
Sigma-Aldrich
Monoclonal Anti-PPP2R1A antibody produced in mouse, clone 4E6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-IQGAP1 Antibody, clone AF4, clone AF4, Upstate®, from mouse
Sigma-Aldrich
Normal Mouse IgG, This Normal Mouse IgG is validated for use in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of Mouse IgG, Non-immune.
Sigma-Aldrich
Anti-DYRK1A Mouse mAb (7D10), liquid, clone 7D10, Calbiochem®
Sigma-Aldrich
Anti-mPER1 (residues 6-21) Antibody, serum, from rabbit