  • Search Results
  • Zerumbone, a Tropical Ginger Sesquiterpene of Zingiber officinale Roscoe, Attenuates α-MSH-Induced Melanogenesis in B16F10 Cells.
Zerumbone, a Tropical Ginger Sesquiterpene of Zingiber officinale Roscoe, Attenuates α-MSH-Induced Melanogenesis in B16F10 Cells.
International journal of molecular sciences (2018-10-17)
Taek-In Oh, Hye-Jeong Jung, Yoon-Mi Lee, Sujin Lee, Geon-Hee Kim, Sang-Yeon Kan, Hyeji Kang, Taerim Oh, Hyun Myung Ko, Keun-Chang Kwak, Ji-Hong Lim
ABSTRACT
Zerumbone (ZER), an active constituent of the Zingiberaceae family, has been shown to exhibit several biological activities, such as anti-inflammatory, anti-allergic, anti-microbial, and anti-cancer; however, it has not been studied for anti-melanogenic properties. In the present study, we demonstrate that ZER and Zingiber officinale (ZO) extract significantly attenuate melanin accumulation in α-melanocyte-stimulating hormone (α-MSH)-stimulated mouse melanogenic B16F10 cells. Further, to elucidate the molecular mechanism by which ZER suppresses melanin accumulation, we analyzed the expression of melanogenesis-associated transcription factor, microphthalmia-associated transcription factor (MITF), and its target genes, such as tyrosinase, tyrosinase-related protein 1 (TYRP1), and tyrosinase-related protein 2 (TYRP2), in B16F10 cells that are stimulated by α-MSH. Here, we found that ZER inhibits the MITF-mediated expression of melanogenic genes upon α-MSH stimulation. Additionally, cells treated with different concentrations of zerumbone and ZO showed increased extracellular signal-regulated kinases 1 and 2 (ERK1/2) phosphorylation, which are involved in the degradation mechanism of MITF. Pharmacological inhibition of ERK1/2 using U0126 sufficiently reversed the anti-melanogenic effect of ZER, suggesting that increased phosphorylation of ERK1/2 is required for its anti-melanogenic activity. Taken together, these results suggest that ZER and ZO extract can be used as active ingredients in skin-whitening cosmetics because of their anti-melanogenic effect.
Sigma-Aldrich
α-Melanocyte stimulating hormone, ≥97% (HPLC)
Sigma-Aldrich
Zerumbone, ≥98% (HPLC)
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
Arbutin, ≥98%
Sigma-Aldrich
L-Dopa-(phenyl-d3), ≥98 atom % D, ≥98% (CP)