HomeSupportVolume Unit Converter
Volume Unit Converter
Volume is defined as the three-dimensional space enclosed by a boundary or occupied by an object. The formula for the volume of various three-dimensional spaces are as follows:
Cube: a3; where a is the length of each side
Rectangle: l × w × h
Sphere: 4/3 π r3
Cylinder: π × r2 × h
Cone: 1/3 π × r2
Use the following calculator to convert a known volume into various units.