
Zanamivir impurity F

Synonym(s):
1H-Pyrazole-1-carboxamidine hydrochloride, 1-Amidinopyrazole hydrochloride, Praxadine
Empirical Formula (Hill Notation):
C4H6N4 · HCl
CAS Number:
Molecular Weight:
146.58
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing