Skip to Content
Merck
vibrant-m

SUPELCOGEL Ca, 6% Crosslinked HPLC Column

vibrant-m
Synonym(s):
SUPELCOGEL 6% Cross Linked HPLC Columns

Products

Shipping
Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Technique
Showing 1-1 of 1