Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Colistimethate Sodium
Synonym(s):
Colistimethate sodium, Colistin sulphomethate sodium
CAS Number:
8068-28-8
EC Number:
232-516-9
Products
Brand
Supelco (1)
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (4)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
PHR1737
Colistimethate Sodium,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
1147009
Colistimethate Sodium,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
Y0001888
Colistimethate Sodium,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
Y0001887
Colistimethate Sodium,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
Y0001886
Colistimethate Sodium,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing