Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Paraffin (White soft)
Synonym(s):
Paraffin wax
CAS Number:
8002-74-2
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Y0000913
Paraffin (White soft),
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
1496904
Paraffin (White soft),
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
Y0000780
Paraffin (White soft),
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
Y0000912
Paraffin (White soft),
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing