Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluoride standard solution
Synonym(s):
Sodium fluoride solution
Linear Formula:
NaF
CAS Number:
7681-49-4
Molecular Weight:
41.99
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (3)
Supelco (2)
Formula Weight
Physical Form
Format
Quality Segment
Application
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.19814
Fluoride standard solution,
traceable to SRM from NIST NaF in H₂O 1000 mg/l F Certipur
®
Expand
View Pricing
919
Fluoride standard solution,
Expand
View Pricing
67414
Fluoride standard solution,
Expand
View Pricing
47072
Fluoride standard solution,
0.1 M F
-
, analytical standard (for ion-selective electrodes)
Expand
View Pricing
11-0475
Fluoride standard solution,
1 mg/mL F
-
Expand
View Pricing